DR2313 97%

Synonyme: 7,8-Dihydro-2-methyl-1H-thiopyrano[4,3-d]pyrimidin-4(5H)-one

BD160273-25MG
BLDPBD160273-25MGEA 272 EUR
BLDPBD160273-25MG
DR2313 97%
DR2313

Storage condition: Sealed in dry, 2-8°C

Formel: C₈H₁₀N₂OS
Molecular Weight: 182,2428 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD08703132
CAS-Nummer: 284028-90-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR