5-Bromopyrazolo[1,5-a]pyrimidine 97%

BD160231-250MG BD160231-5G BD160231-100MG BD160231-1G
BLDPBD160231-250MGEA 48 EUR
BLDPBD160231-250MG BLDPBD160231-5G BLDPBD160231-100MG BLDPBD160231-1G
5-Bromopyrazolo[1,5-a]pyrimidine 97%
5-Bromopyrazolo[1,5-a]pyrimidine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₄BrN₃
Molecular Weight: 198,02 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD11042744
CAS-Nummer: 1159981-95-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR