Methyl-2,3-dihydroxybenzoat 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD160143-100GEA 397 EUR
BLDPBD160143-100G BLDPBD160143-10G BLDPBD160143-5G BLDPBD160143-25G BLDPBD160143-1G
Methyl-2,3-dihydroxybenzoat 98%
Methyl-2,3-dihydroxybenzoat

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₈O₄
Molecular Weight: 168,1467 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00039520
CAS-Nummer: 2411-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR