5-Methyl-3-nitro-1H-pyrazole 95%

BD160131-5G BD160131-1G BD160131-100G BD160131-25G BD160131-10G
BLDPBD160131-5GEA 25 EUR
BLDPBD160131-5G BLDPBD160131-1G BLDPBD160131-100G BLDPBD160131-25G BLDPBD160131-10G
5-Methyl-3-nitro-1H-pyrazole 95%
5-Methyl-3-nitro-1H-pyrazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₄H₅N₃O₂
Molecular Weight: 127,1 g/mol
Lagertemperatur: Ambient
MDL: MFCD00514181
CAS-Nummer: 1048925-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR