1-Brom-3-(difluormethyl)benzol 98%

Synonyme: 1-Bromo-3-(difluoromethyl)benzene,3-Bromobenzal fluoride

BD160129-1G BD160129-5G BD160129-25G BD160129-10G
BLDPBD160129-1GEA 13 EUR
BLDPBD160129-1G BLDPBD160129-5G BLDPBD160129-25G BLDPBD160129-10G
1-Brom-3-(difluormethyl)benzol 98%
1-Bromo-3-(difluoromethyl)benzene

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₅BrF₂
Molecular Weight: 207,0154 g/mol
Lagertemperatur: Ambient
MDL: MFCD07368984
CAS-Nummer: 29848-59-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR