5-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid 98%

BLDPBD160123-250MGEA 29 EUR
BLDPBD160123-250MG BLDPBD160123-1G BLDPBD160123-5G
5-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid 98%
5-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₆H₆BrNO₂
Molecular Weight: 204,0213 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD16659627
CAS-Nummer: 865186-82-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR