4,5-Dichloropyrimidin-2-amine 98%

BLDPBD160093-10GEA 447 EUR
BLDPBD160093-10G BLDPBD160093-5G BLDPBD160093-250MG BLDPBD160093-1G BLDPBD160093-100MG BLDPBD160093-25G
4,5-Dichloropyrimidin-2-amine 98%
4,5-Dichloropyrimidin-2-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₄H₃Cl₂N₃
Molecular Weight: 163,9900 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13193667
CAS-Nummer: 403854-21-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR