7-Methoxy-4-oxo-3,4-dihydroquinazolin-6-yl acetate 97%

BD160075-10G BD160075-25G BD160075-100G BD160075-5G BD160075-1G
BLDPBD160075-10GEA 42 EUR
BLDPBD160075-10G BLDPBD160075-25G BLDPBD160075-100G BLDPBD160075-5G BLDPBD160075-1G
7-Methoxy-4-oxo-3,4-dihydroquinazolin-6-yl acetate 97%
7-Methoxy-4-oxo-3,4-dihydroquinazolin-6-yl acetate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₁H₁₀N₂O₄
Molecular Weight: 234,21 g/mol
Lagertemperatur: Ambient
MDL: MFCD09833010
CAS-Nummer: 179688-53-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR