Ethyl 2-Oxocycloheptanecarboxylate 95%

Synonyme: 2-Oxocycloheptanecarboxylic acid Ethyl ester

BD160057-100MG
BLDPBD160057-100MGEA 57 EUR
BLDPBD160057-100MG
Ethyl 2-Oxocycloheptanecarboxylate 95%
Ethyl 2-Oxocycloheptanecarboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₀H₁₆O₃
Molecular Weight: 184,24 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00604086
CAS-Nummer: 774-05-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR