2,3,4,5-Tetrahydro-1H-benzo[c]azepine hydrochloride 97%

BD160050-1G BD160050-100MG BD160050-250MG
BLDPBD160050-1GEA 95 EUR
BLDPBD160050-1G BLDPBD160050-100MG BLDPBD160050-250MG
2,3,4,5-Tetrahydro-1H-benzo[c]azepine hydrochloride 97%
2,3,4,5-Tetrahydro-1H-benzo[c]azepine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₀H₁₄ClN
Molecular Weight: 183,6800 g/mol
Lagertemperatur: Ambient
MDL: MFCD12198423
CAS-Nummer: 17724-36-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR