Tetrahydrothiopyran-4-carbonitrile 98%

BD160020-250MG BD160020-1G BD160020-5G
BLDPBD160020-250MGEA 19 EUR
BLDPBD160020-250MG BLDPBD160020-1G BLDPBD160020-5G
Tetrahydrothiopyran-4-carbonitrile 98%
Tetrahydrothiopyran-4-carbonitrile

Storage condition: Sealed in dry, 2-8°C

Formel: C₆H₉NS
Molecular Weight: 127,21 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09025909
CAS-Nummer: 195503-40-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR