Ethyl-5-bromopyrimidine-2-carboxylate 95%

BLDPBD160005-5GEA 92 EUR
BLDPBD160005-5G BLDPBD160005-250MG BLDPBD160005-25G BLDPBD160005-1G
Ethyl-5-bromopyrimidine-2-carboxylate 95%
Ethyl-5-bromopyrimidine-2-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₇BrN₂O₂
Molecular Weight: 231,05 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1197193-30-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR