(R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-4-hydroxybenzeneacetic acid 96%

Synonyme: (R)-2-(4-Ethyl-2,3-dioxopiperazine-1-carboxamido)-2-(4-hydroxyphenyl)acetic acid,(R)-(-)-2-(4-Ethyl-2,3-dioxopiperazine-1-carboxamido)-2-(4-hydroxyphenyl)acetic acid

BD15998-1G BD15998-250MG BD15998-100G BD15998-500G BD15998-10G BD15998-25G BD15998-5G
BLDPBD15998-1GEA 13 EUR
BLDPBD15998-1G BLDPBD15998-250MG BLDPBD15998-100G BLDPBD15998-500G BLDPBD15998-10G BLDPBD15998-25G BLDPBD15998-5G
(R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-4-hydroxybenzeneacetic acid 96%
(R)-(-)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-4-hydroxybenzeneacetic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₅H₁₇N₃O₆
Molecular Weight: 335,32 g/mol
Lagertemperatur: Ambient
MDL: MFCD00792472
CAS-Nummer: 62893-24-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR