3,3-Diphenylpropylamin 98%

Synonyme: 3,3-Diphenylpropan-1-amine

BD15996-1G BD15996-100G BD15996-5G BD15996-25G
BLDPBD15996-1GEA 9 EUR
BLDPBD15996-1G BLDPBD15996-100G BLDPBD15996-5G BLDPBD15996-25G
3,3-Diphenylpropylamin 98%
3,3-Diphenylpropylamin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₅H₁₇N
Molecular Weight: 211,31 g/mol
Lagertemperatur: Ambient
MDL: MFCD00008202
CAS-Nummer: 5586-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR