(S)-tert-Butyl 2-cyanomorpholine-4-carboxylate 95%

BD159957-100MG BD159957-250MG
BLDPBD159957-100MGEA 105 EUR
BLDPBD159957-100MG BLDPBD159957-250MG
(S)-tert-Butyl 2-cyanomorpholine-4-carboxylate 95%
(S)-tert-Butyl 2-cyanomorpholine-4-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₀H₁₆N₂O₃
Molecular Weight: 212,24 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD16659643
CAS-Nummer: 1257856-31-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR