(R)-4-Boc-2-(aminomethyl)morpholine 97%

Synonyme: (R)-tert-Butyl 2-(aminomethyl)morpholine-4-carboxylate

BD159956-5G BD159956-1G BD159956-25G BD159956-250MG
BLDPBD159956-5GEA 338 EUR
BLDPBD159956-5G BLDPBD159956-1G BLDPBD159956-25G BLDPBD159956-250MG
(R)-4-Boc-2-(aminomethyl)morpholine 97%
(R)-4-Boc-2-(aminomethyl)morpholine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₀H₂₀N₂O₃
Molecular Weight: 216,277 g/mol
Lagertemperatur: Ambient
MDL: MFCD13189480
CAS-Nummer: 1174913-80-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR