(S)-2-(N-Boc-aminomethyl)morpholine 95%

BLDPBD159946-1GEA 517 EUR
BLDPBD159946-1G BLDPBD159946-100MG BLDPBD159946-250MG
(S)-2-(N-Boc-aminomethyl)morpholine 95%
(S)-2-(N-Boc-aminomethyl)morpholine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₀H₂₀N₂O₃
Molecular Weight: 216,277 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12545849
CAS-Nummer: 875551-59-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR