2-Difluoromethyl-1H-benzoimidazole 98%

BLDPBD159938-100GEA 688 EUR
BLDPBD159938-100G BLDPBD159938-5G BLDPBD159938-1G BLDPBD159938-250MG BLDPBD159938-10G BLDPBD159938-25G
2-Difluoromethyl-1H-benzoimidazole 98%
2-Difluoromethyl-1H-benzoimidazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆F₂N₂
Molecular Weight: 168,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00463015
CAS-Nummer: 705-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR