2,3-Dihydro-1H-pyrrolo[3,4-c]pyridine dihydrochloride 97%

BD159935-1G BD159935-25G BD159935-250MG BD159935-10G BD159935-5G
BLDPBD159935-1GEA 136 EUR
BLDPBD159935-1G BLDPBD159935-25G BLDPBD159935-250MG BLDPBD159935-10G BLDPBD159935-5G
2,3-Dihydro-1H-pyrrolo[3,4-c]pyridine dihydrochloride 97%
2,3-Dihydro-1H-pyrrolo[3,4-c]pyridine dihydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₈N₂·2ClH
Molecular Weight: 193,074 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD15144657
CAS-Nummer: 6000-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR