2-Oxa-6-azaspiro[3.3]heptane 97%

BLDPBD159921-1GEA 73 EUR
BLDPBD159921-1G BLDPBD159921-25G BLDPBD159921-250MG BLDPBD159921-10G BLDPBD159921-100MG BLDPBD159921-5G
2-Oxa-6-azaspiro[3.3]heptane 97%
2-Oxa-6-azaspiro[3.3]heptane
Formel: C₅H₉NO
MDL: MFCD12407093
CAS-Nummer: 174-78-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR