(R)-4-Benzyl-2-hydroxymethylpiperazine 98%

BD159917-100MG BD159917-250MG BD159917-1G
BLDPBD159917-100MGEA 134 EUR
BLDPBD159917-100MG BLDPBD159917-250MG BLDPBD159917-1G
(R)-4-Benzyl-2-hydroxymethylpiperazine 98%
(R)-4-Benzyl-2-hydroxymethylpiperazine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₂H₁₈N₂O
Molecular Weight: 206,284 g/mol
Lagertemperatur: Ambient
MDL: MFCD11113046
CAS-Nummer: 149715-46-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR