(R)-tert-Butyl 2-(2-hydroxyethyl)morpholine-4-carboxylate 97%

BD159915-100MG BD159915-250MG BD159915-50MG
BLDPBD159915-100MGEA 162 EUR
BLDPBD159915-100MG BLDPBD159915-250MG BLDPBD159915-50MG
(R)-tert-Butyl 2-(2-hydroxyethyl)morpholine-4-carboxylate 97%
(R)-tert-Butyl 2-(2-hydroxyethyl)morpholine-4-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₁H₂₁NO₄
Molecular Weight: 231,29 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD11865208
CAS-Nummer: 136992-21-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR