3-Carboxy-4-chlorphenylboronsäure 98%

Synonyme: 5-Borono-2-chlorobenzoic acid

BD159891-100MG BD159891-5G BD159891-25G BD159891-10G BD159891-1G BD159891-250MG
BLDPBD159891-100MGEA 6 EUR
BLDPBD159891-100MG BLDPBD159891-5G BLDPBD159891-25G BLDPBD159891-10G BLDPBD159891-1G BLDPBD159891-250MG
3-Carboxy-4-chlorphenylboronsäure 98%
3-Carboxy-4-chlorphenylboronsäure

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₆BClO₄
Molecular Weight: 200,3800 g/mol
Lagertemperatur: Ambient
MDL: MFCD06801689
CAS-Nummer: 913835-32-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR