4-Acetoxymethylbenzeneboronic acid 98%

Synonyme: (4-(Acetoxymethyl)phenyl)boronic acid

BD159882-1G BD159882-5G BD159882-250MG
BLDPBD159882-1GEA 113 EUR
BLDPBD159882-1G BLDPBD159882-5G BLDPBD159882-250MG
4-Acetoxymethylbenzeneboronic acid 98%
4-Acetoxymethylbenzeneboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₁₁BO₄
Molecular Weight: 193,99 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD08235082
CAS-Nummer: 326496-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR