2-Amino-6-methoxybenzonitrile 98%

BD159814-5G BD159814-1G BD159814-250MG BD159814-25G
BLDPBD159814-5GEA 132 EUR
BLDPBD159814-5G BLDPBD159814-1G BLDPBD159814-250MG BLDPBD159814-25G
2-Amino-6-methoxybenzonitrile 98%
2-Amino-6-methoxybenzonitrile

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₈N₂O
Molecular Weight: 148,162 g/mol
Lagertemperatur: Ambient
MDL: MFCD09745275
CAS-Nummer: 1591-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR