tert-Butyl-3-oxocyclobutanecarboxylate 95%

BLDPBD159804-10GEA 64 EUR
BLDPBD159804-10G BLDPBD159804-5G BLDPBD159804-25G BLDPBD159804-1G BLDPBD159804-100G
tert-Butyl-3-oxocyclobutanecarboxylate 95%
tert-Butyl-3-oxocyclobutanecarboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₄O₃
Molecular Weight: 170,208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12827541
CAS-Nummer: 145549-76-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR