6-Amino-3-bromo-2,4-dimethylpyridine 98%

BLDPBD159799-5GEA 280 EUR
BLDPBD159799-5G BLDPBD159799-250MG BLDPBD159799-25G BLDPBD159799-1G
6-Amino-3-bromo-2,4-dimethylpyridine 98%
6-Amino-3-bromo-2,4-dimethylpyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₉BrN₂
Molecular Weight: 201,064 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00151793
CAS-Nummer: 89856-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR