4-Cyano-5-bromopyrimidine 95%

Synonyme: 5-Bromo-4-cyanopyrimidine

BD159796-100MG BD159796-1G BD159796-5G BD159796-250MG
BLDPBD159796-100MGEA 72 EUR
BLDPBD159796-100MG BLDPBD159796-1G BLDPBD159796-5G BLDPBD159796-250MG
4-Cyano-5-bromopyrimidine 95%
4-Cyano-5-bromopyrimidine

Storage condition: Sealed in dry, 2-8°C

Formel: C₅H₂BrN₃
Molecular Weight: 184 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09999211
CAS-Nummer: 114969-66-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR