Boc-(S)-3-amino-4-pentafluoro-phenylbutyric acid 98%

Synonyme: (S)-3-((tert-Butoxycarbonyl)amino)-4-(perfluorophenyl)butanoic acid

BD15977-1G BD15977-100MG BD15977-250MG
BLDPBD15977-1GEA 498 EUR
BLDPBD15977-1G BLDPBD15977-100MG BLDPBD15977-250MG
Boc-(S)-3-amino-4-pentafluoro-phenylbutyric acid 98%
Boc-(S)-3-amino-4-pentafluoro-phenylbutyric acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₅H₁₆F₅NO₄
Molecular Weight: 369,29 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD01861038
CAS-Nummer: 270063-42-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR