Boc-Phe-Phe-OH

Lieferant: BLD PHARMATECH GMBH
BD1597-1G BD1597-25G BD1597-5G BD1597-250MG
BLDPBD1597-1GEA 14 EUR
BLDPBD1597-1G BLDPBD1597-25G BLDPBD1597-5G BLDPBD1597-250MG
Boc-Phe-Phe-OH
Proteine und Peptide
  • GHS07

Storage condition: Sealed in dry, 2-8°C
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR