2,6-Difluor-4-methoxyanilin 98%

Synonyme: 4-Amino-3,5-difluoranisol

BD159691-10G BD159691-5G BD159691-250MG BD159691-25G BD159691-1G
BLDPBD159691-10GEA 229 EUR
BLDPBD159691-10G BLDPBD159691-5G BLDPBD159691-250MG BLDPBD159691-25G BLDPBD159691-1G
2,6-Difluor-4-methoxyanilin 98%
2,6-Difluor-4-methoxyanilin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₇F₂NO
Lagertemperatur: Ambient
MDL: MFCD04115922
CAS-Nummer: 151414-47-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR