6-Morpholinopyrazin-2-amine 97%

BD159673-1G BD159673-100MG BD159673-250MG
BLDPBD159673-1GEA 78 EUR
BLDPBD159673-1G BLDPBD159673-100MG BLDPBD159673-250MG
6-Morpholinopyrazin-2-amine 97%
6-Morpholinopyrazin-2-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₈H₁₂N₄O
Molecular Weight: 180,2100 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD05722015
CAS-Nummer: 717847-03-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR