Methyl-5-brom-6-hydroxynicotinat 97%

Synonyme: Methyl 5-bromo-6-hydroxynicotinate

BD159671-250MG BD159671-5G BD159671-1G BD159671-25G BD159671-10G
BLDPBD159671-250MGEA 18 EUR
BLDPBD159671-250MG BLDPBD159671-5G BLDPBD159671-1G BLDPBD159671-25G BLDPBD159671-10G
Methyl-5-brom-6-hydroxynicotinat 97%
Methyl-5-brom-6-hydroxynicotinat

Storage condition: Sealed in dry, 2-8°C

Formel: C₇H₆BrNO₃
Molecular Weight: 232,03 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD08235247
CAS-Nummer: 381247-99-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR