6-Chloro-N4-propylpyrimidine-4,5-diamine 98%

BD159650-5G BD159650-1G BD159650-250MG
BLDPBD159650-5GEA 487 EUR
BLDPBD159650-5G BLDPBD159650-1G BLDPBD159650-250MG
6-Chloro-N4-propylpyrimidine-4,5-diamine 98%
6-Chloro-N4-propylpyrimidine-4,5-diamine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₇H₁₁ClN₄
Molecular Weight: 186,6420 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD07440144
CAS-Nummer: 195252-56-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR