6-Methylpyrimidin-4-amine 97%

BLDPBD159647-250MGEA 79 EUR
BLDPBD159647-250MG BLDPBD159647-100MG BLDPBD159647-5G BLDPBD159647-1G
6-Methylpyrimidin-4-amine 97%
6-Methylpyrimidin-4-amine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₅H₇N₃
Molecular Weight: 109,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00188073
CAS-Nummer: 3435-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR