5-Bromo-2-fluoronicotinaldehyde 98%

BLDPBD159627-500GEA 1941 EUR
BLDPBD159627-500G BLDPBD159627-100G BLDPBD159627-25G BLDPBD159627-1G BLDPBD159627-10G BLDPBD159627-5G
5-Bromo-2-fluoronicotinaldehyde 98%
5-Bromo-2-fluoronicotinaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₃BrFNO
Molecular Weight: 204,00 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09842440
CAS-Nummer: 875781-15-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR