7-Bromo-1H-pyrrolo[3,2-d]pyrimidin-4(5H)-one 98%

BD159623-100MG BD159623-1G BD159623-250MG
BLDPBD159623-100MGEA 105 EUR
BLDPBD159623-100MG BLDPBD159623-1G BLDPBD159623-250MG
7-Bromo-1H-pyrrolo[3,2-d]pyrimidin-4(5H)-one 98%
7-Bromo-1H-pyrrolo[3,2-d]pyrimidin-4(5H)-one

Storage condition: Sealed in dry, 2-8°C

Formel: C₆H₄BrN₃O
Molecular Weight: 214,0195 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09999206
CAS-Nummer: 93587-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR