tert-Butyl-5-bromo-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate 95%

BLDPBD159593-5GEA 601 EUR
BLDPBD159593-5G BLDPBD159593-250MG BLDPBD159593-100MG BLDPBD159593-1G BLDPBD159593-10G BLDPBD159593-25G
tert-Butyl-5-bromo-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate 95%
tert-Butyl-5-bromo-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₇H₂₁BrN₂O₃
Molecular Weight: 381,2600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10566052
CAS-Nummer: 873779-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR