3-Benzyl-7-chloro-3H-[1,2,3]triazolo[4,5-d]pyrimidine 98%

BD159591-5G BD159591-250MG BD159591-1G BD159591-100MG
BLDPBD159591-5GEA 664 EUR
BLDPBD159591-5G BLDPBD159591-250MG BLDPBD159591-1G BLDPBD159591-100MG
3-Benzyl-7-chloro-3H-[1,2,3]triazolo[4,5-d]pyrimidine 98%
3-Benzyl-7-chloro-3H-[1,2,3]triazolo[4,5-d]pyrimidine

Storage condition: Inert atmosphere, 2-8°C

Formel: C11H8ClN5
Molecular Weight: 245,6700 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09038669
CAS-Nummer: 21410-06-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR