3,6-Difluoropyridazine 98%

BLDPBD159585-1GEA 49 EUR
BLDPBD159585-1G BLDPBD159585-5G BLDPBD159585-100MG BLDPBD159585-250MG
3,6-Difluoropyridazine 98%
3,6-Difluoropyridazine

Storage condition: Inert atmosphere, Room Temperature

Formel: C4H2F2N2
Molecular Weight: 116,0700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09909693
CAS-Nummer: 33097-39-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR