Ethyl-3-aminofuro[2,3-b]pyridine-2-carboxylate 95%

Synonyme: Ethyl 3-aminofuro[2,3-b]pyridine-2-carboxylate

BD159580-1G BD159580-100MG BD159580-250MG
BLDPBD159580-1GEA 111 EUR
BLDPBD159580-1G BLDPBD159580-100MG BLDPBD159580-250MG
Ethyl-3-aminofuro[2,3-b]pyridine-2-carboxylate 95%
Ethyl-3-aminofuro[2,3-b]pyridine-2-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₀H₁₀N₂O₃
Molecular Weight: 206,1980 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09837639
CAS-Nummer: 371945-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR