Ethyl-2-(3,5-dichlorophenyl)thiazole-4-carboxylate 98%

Synonyme: Ethyl 2-(3,5-dichlorophenyl)thiazole-4-carboxylate

BD159557-250MG
BLDPBD159557-250MGEA 10 EUR
BLDPBD159557-250MG
Ethyl-2-(3,5-dichlorophenyl)thiazole-4-carboxylate 98%
Ethyl-2-(3,5-dichlorophenyl)thiazole-4-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₉Cl₂NO₂S
Molecular Weight: 302,18 g/mol
Lagertemperatur: Ambient
MDL: MFCD09743714
CAS-Nummer: 132089-38-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR