(3-Bromoisoxazol-5-yl)methylamine hydrochloride 97%

BLDPBD159528-250MGEA 37 EUR
BLDPBD159528-250MG BLDPBD159528-5G BLDPBD159528-1G
(3-Bromoisoxazol-5-yl)methylamine hydrochloride 97%
(3-Bromoisoxazol-5-yl)methylamine hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₄H₆BrClN₂O
Molecular Weight: 213,47 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08705903
CAS-Nummer: 90802-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR