2-(2-Fluorophenyl)pyrimidine-5-carbaldehyde 98%

BD159508-250MG BD159508-1G
BLDPBD159508-250MGEA 55 EUR
BLDPBD159508-250MG BLDPBD159508-1G
2-(2-Fluorophenyl)pyrimidine-5-carbaldehyde 98%
2-(2-Fluorophenyl)pyrimidine-5-carbaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₁H₇FN₂O
Molecular Weight: 202,1800 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09743685
CAS-Nummer: 946707-17-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR