4-Brom-4'-propylbiphenyl 98%

BLDPBD15948-100GEA 121 EUR
BLDPBD15948-100G BLDPBD15948-5G BLDPBD15948-25G
4-Brom-4'-propylbiphenyl 98%
4-Brom-4'-propylbiphenyl

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₅H₁₅Br
Molecular Weight: 275,1800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00060105
CAS-Nummer: 58743-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR