tert-Butyl-5-bromo-3-(hydroxymethyl)-1H-indole-1-carboxylate 98%

Synonyme: 5-Bromo-3-(hydroxymethyl)-1H-indole, N-BOC protected,tert-Butyl 5-bromo-3-(hydroxymethyl)-1H-indole-1-carboxylate

BD159457-250MG BD159457-100MG BD159457-1G BD159457-5G
BLDPBD159457-250MGEA 13 EUR
BLDPBD159457-250MG BLDPBD159457-100MG BLDPBD159457-1G BLDPBD159457-5G
tert-Butyl-5-bromo-3-(hydroxymethyl)-1H-indole-1-carboxylate 98%
tert-Butyl-5-bromo-3-(hydroxymethyl)-1H-indole-1-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₄H₁₆BrNO₃
Molecular Weight: 326,1857 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD05864710
CAS-Nummer: 905710-14-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR