Methyl-2-amino-5-chlorothiazole-4-carboxylate 95%

Synonyme: Methyl 2-amino-5-chlorothiazole-4-carboxylate

BD159427-250MG BD159427-1G
BLDPBD159427-250MGEA 8 EUR
BLDPBD159427-250MG BLDPBD159427-1G
Methyl-2-amino-5-chlorothiazole-4-carboxylate 95%
Methyl-2-amino-5-chlorothiazole-4-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₅H₅ClN₂O₂S
Molecular Weight: 192,6200 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD05864655
CAS-Nummer: 914348-76-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR