2-Methylimidazo[4,5-c]pyridin 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD159414-250MGEA 49 EUR
BLDPBD159414-250MG BLDPBD159414-1G
2-Methylimidazo[4,5-c]pyridin 95%
2-Methylimidazo[4,5-c]pyridin

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₇N₃
Molecular Weight: 133,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03426025
CAS-Nummer: 63604-59-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR