3,5-Dichlor-4-methoxyphenylboronsäure 98%

Synonyme: (3,5-Dichloro-4-methoxyphenyl)boronic acid

BD159398-250MG BD159398-1G BD159398-5G
BLDPBD159398-250MGEA 85 EUR
BLDPBD159398-250MG BLDPBD159398-1G BLDPBD159398-5G
3,5-Dichlor-4-methoxyphenylboronsäure 98%
3,5-Dichlor-4-methoxyphenylboronsäure

Storage condition: Sealed in dry, 2-8°C

Formel: C₇H₇BCl₂O₃
Molecular Weight: 220,8500 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD04039018
CAS-Nummer: 175883-61-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR