5-Bromo-1,3-di-tert-butyl-2-methoxybenzene 98%

BD159391-25G BD159391-1G BD159391-250MG BD159391-100G BD159391-5G BD159391-10G
BLDPBD159391-25GEA 451 EUR
BLDPBD159391-25G BLDPBD159391-1G BLDPBD159391-250MG BLDPBD159391-100G BLDPBD159391-5G BLDPBD159391-10G
5-Bromo-1,3-di-tert-butyl-2-methoxybenzene 98%
5-Bromo-1,3-di-tert-butyl-2-methoxybenzene

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₅H₂₃BrO
Molecular Weight: 299,2500 g/mol
Lagertemperatur: Ambient
MDL: MFCD12024327
CAS-Nummer: 1516-96-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR